Learn More
Indofine Chemical Baicalin , 99%, 1 Gm
SDP
Supplier:  Indofine Chemical 06012 1 GM
Encompass_Preferred
BAICALIN 1 GM, Flavone, Scutellaria baicalensis; Baicalein 7-d-glucuronide; B-d-gluopyranosiduronic acid-5,6-dihydroxy-4-oxo-2-phenyl-4h-1-benzopyran-7-yl, C21H18O11, MW: 446.36, 0.99, 21967-41-9, MFCD00134418
Catalog No. 50-908-348