Learn More
Cole-Parmer Bain Marie Container Snap-Tight, Positive-Lock Lid, HDPE, for 63530-10, -12, -14
Supplier:  Cole-Parmer 6353053
Encompass_Preferred
Snap-tight, positive-lock lids are made of FDA compliant High-Density PolyEthylene (HDPE) and can be frozen, boiled, or chemically sterilized. Bain Marie HDPE containers are sold separately.
Catalog No. NC4001331