Learn More
Cole-Parmer Bain Marie HDPE Container, 2 quart; 12/Pk
Supplier:  Cole-Parmer 6353000
Encompass_Preferred
Containers are made of FDA-compliant HDPE and can be frozen, boiled, or chemically sterilized. Snap-tight, positive-lock lids are sold separately. NSF certified.
Catalog No. NC0986823