Learn More
Cayman Chemical Bak BH372-87humntrIf 10mg
SDP
Supplier:  Cayman Chemical 3690910 MG
Encompass_Preferred
A Bak-derived peptide; selectively binds to Bcl-xL over Bcl-2 (EC50s = 174 and 3,020 nM, respectively); induces apoptosis in LLC-PK1 cells
Catalog No. 50-291-5051