Learn More
Selleck Chemical LLC BAL-0028 E1970-5MG
SDP
Supplier:  Selleck Chemical LLC E19705MG
Encompass_Preferred
BAL-0028 (Compound 3) which belongs to the indazole class is an inhibitor of NLRP3 activation with an IC50 value of 25 nM BAL-0028 binds tightly to the NACHT domain of NLRP3 with KD values ranging from 104 to 123 nM BAL-0028 also exhibits anti-inflammatory activity
Catalog No. 50-000-39482