Learn More
Medchemexpress LLC BAL-0028 | 2842012-69-3 | 99.9% | 403.45 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY162333
Encompass_Preferred
BAL-0028 is a reversible inhibitor of NLRP3 activation. It binds closely to the NACHT domain of the NLRP3, inhibiting the secretion of IL-1β, and demonstrating anti-inflammatory activity.
  • Reversible inhibitor of NLRP3 activation
  • Binds to the NACHT domain of NLRP3
  • Inhibits secretion of IL-1β
  • Has anti-inflammatory activity
Catalog No. 50-003-95218