Learn More
Medchemexpress LLC Baliforsen (sodium) | 1687746-79-7 | 5616 (free acid) | 5 MG

Supplier: Medchemexpress LLC HY1457255MG
Baliforsen (sodium) is a 16-nucleotide antisense oligonucleotide that binds myotonic dystrophy protein kinase (DMPK) mRNA for use in laboratory research into myotonic dystrophy. Supplied as the sodium salt, the compound is intended for research use only and is not validated for medical applications.
- Targets DMPK mRNA to reduce transcript levels.
- 16-nucleotide length with modified 2'-O-(2-methoxyethyl) and phosphorothioate linkages.
- Sodium salt form for improved handling and solubility.
- Molecular weight approximately 5,616 (free acid).
- Store at -20°C sealed; in solvent keep at -80°C up to 6 months or -20°C up to 1 month.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.