Learn More
Medchemexpress LLC Balipodect | 1238697-26-1 | MFCD28385852 | ≥98.0% | 428.4 g/mol | C23H17FN6O2 | 50MG
SDP
Supplier:  Medchemexpress LLC HY1247250MG
Encompass_Preferred
Balipodect (TAK-063) is a potent, selective, orally active phosphodiesterase 10A (PDE10A) inhibitor used in research to investigate PDE10A-mediated signaling and neuropsychiatric mechanisms. It demonstrates subnanomolar potency and high selectivity, and is supplied for research use only.
  • Potent PDE10A inhibition with IC50 0.30 nM
  • High selectivity (>15,000-fold) for reduced off-target activity
  • Orally active small molecule suitable for in vivo and in vitro studies
  • High purity (≥98.0%) for analytical and experimental consistency
  • Defined chemical identity: C23H17FN6O2, MW 428.4 g/mol, CAS 1238697-26-1
Catalog No. 50-002-35877