Learn More
Medchemexpress LLC HY-P1682A 5mg Medchemexpress, Balixafortide (TFA) CAS:0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP1682A5MG
Encompass_Preferred
Medchemexpress, HY-P1682A 5mg Balixafortide (TFA) CAS:0 Balixafortide TFA (POL6326 TFA) is a potent, selective, well-tolerated peptidic CXCR4 antagonist with an IC50 < 10 nM. Balixafortide TFA shows 1000-fold selective for CXCR4 than a large panel of receptors including CXCR7. Balixafortide TFA blocks β-arrestin recruitment and calcium flux with IC50s < 10 nM. Balixafortide TFA is also a potent hematopoietic stem and progenitor cell (HSPC) mobilizing agent. Anti-cancer effects[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-1696