Learn More
Grainger Ball End Mill Single End 0.30mm Carbide
Supplier:  Grainger 14K318
Encompass_Preferred
The Grainger Ball End Mill Single End 0.30mm Carbide is a high-performance milling tool designed for precision machining applications. This carbide end mill features a ball nose design for smooth milling of contoured surfaces, and its small 0.30mm diameter allows milling of fine details and small cavities. With a shank diameter of 3mm, this end mill is well-suited for use in mini mills and small CNC machines. The tool's carbide construction provides hardness, wear resistance and strength for long life under demanding conditions. Overall, this is a high-quality ball end mill for fine detail machining in metals, plastics and other materials.
Catalog No. 50-275-0046