Learn More
Grainger Ball End Mill Single End 0.50mm Carbide
Supplier:  Grainger 14M533
Encompass_Preferred
The Micro 100 Ball End Mill Carbide is a 2-flute ball end mill made of solid carbide with a TiAlN coating. It has a 1/4" shank diameter, 1/2" cutting diameter, and 2" overall length. This end mill is designed for general purpose milling in most material types up to 54 Rc hardness. It provides good chip flow in light to medium milling applications. The TiAlN coating helps resist built-up edge and increases tool life in high heat applications. This is a high quality carbide end mill suitable for a wide range of milling operations.
Catalog No. 50-275-1941