Learn More
Grainger Ball End Mill Single End 0.80mm Carbide
Supplier:  Grainger 14M547
Encompass_Preferred
The Ball End Mill from Ball is a high-speed steel end mill with a ball nose end for milling contoured surfaces. It has a 1/4" shank diameter, a 1/2" cutting diameter, and a 2" overall length. The end mill has a TiAlN coating for increased tool life and heat resistance. It is designed for general purpose milling in most material types up to 45 HRC hardness. The end mill provides accurate radii while eliminating sharp corners when profiling.
Catalog No. 50-275-1488