Learn More
Grainger Ball End Mill Single End 0.80mm Carbide
Supplier:  Grainger 14K342
Encompass_Preferred
The Ball End Mill Carbide from Ball is a high-performance end mill for milling a variety of materials. It features a carbide construction for wear resistance and rigidity. The ball end geometry allows for milling of 3D contoured shapes. It has a shank diameter of 1/2 inches, overall length of 4 inches, and a ball diameter of 1/4 inches. The number of flutes is 2. This versatile end mill is well-suited for slotting, profiling, contouring and milling complex 3D shapes.
Catalog No. 50-275-0249