Learn More
Grainger Ball End Mill Single End 1.20mm Carbide
Supplier:  Grainger 14K367
Encompass_Preferred
The Grainger 14K367 is a 14 gauge, type 1 solid copper building wire with THHN/THWN insulation rated for temperatures up to 90°C. This 500 foot spool of wire has a voltage rating of 600V and is UL listed and CSA certified. The wire has a stranded conductor and a minimum bend radius of 4 times the cable diameter. It can be used for general purpose building wire applications including branch circuits, feeders, and services. This American-made wire meets Federal Specification A-A-59544 and complies with NEC article 310.
Catalog No. 50-275-1237