Learn More
Grainger Ball End Mill Single End 1.50mm Carbide
Supplier:  Grainger 14K371
Encompass_Preferred
The Ball End Mill from Ball is a high-speed steel end mill with a ball nose end. It has a 1/2" shank diameter, 1" overall length, and a 1/2" cutting diameter. The end mill has 2 flutes and a right-hand spiral. It is designed for general purpose milling in materials like aluminum, brass, bronze, and copper. The end mill can be run at speeds up to 2000 SFM.
Catalog No. 50-275-1295