Learn More
Grainger Ball End Mill Single End 10.00mm Carbide
Supplier:  Grainger 14K799
Encompass_Preferred
The Ball End Mill from Ball is a high-speed steel end mill with a ball nose end. It has a 1/2" shank diameter, a 2" overall length, and a 1/2" cutting diameter. The end mill has a TiAlN coating for increased tool life and heat resistance. It is designed for general purpose milling in steels, stainless steels, cast irons, aluminum, and other materials. The ball nose geometry allows milling of 3D contoured surfaces. This is a quality end mill made in the USA by Ball, a leading manufacturer of cutting tools.
Catalog No. 50-274-9502