Learn More
Grainger Ball End Mill Single End 2.00mm Carbide
Supplier:  Grainger 14M574
Encompass_Preferred
The Micro 100 Ball End Mill Carbide is a 2-flute ball end mill made of solid carbide with a 1/4" shank diameter, 1/2" cutting diameter, and 2" overall length. It has a TiAIN coating and can be run at speeds up to 15,000 RPM with feed rates up to 80 IPM. This versatile end mill is well-suited for machining contoured surfaces in hardened steels up to 45 Rc. It can produce fine finishes and complex geometries in a wide range of materials. The carbide construction provides wear resistance and tool life. Overall, this is a high quality ball end mill for milling precision parts.
Catalog No. 50-274-8797