Learn More
Grainger Ball End Mill Single End 3.00mm Carbide
Supplier:  Grainger 14K736
Encompass_Preferred
The Ball End Mill from Ball is a high-speed steel end mill with a ball nose end. It has a 1/4" shank diameter, a 1/2" cutting diameter, and a 1-1/2" overall length. The end mill has a TiAlN coating for increased tool life and heat resistance. It is designed for general purpose milling in materials like steel, stainless steel, cast iron, aluminum, copper, brass, and plastics. The ball nose end allows milling of 3D contoured surfaces.
Catalog No. 50-275-1818