Learn More
Grainger Ball End Mill Single End 3/16 Carbide
Supplier:  Grainger 14K672
Encompass_Preferred
The Grainger Approved Ball End Mill is a high-speed steel end mill with a ball nose end. It has a 1/2" shank diameter, 1/2" cutting diameter, 2" overall length, and 2 flute design. This end mill is designed for general purpose milling in most material types up to 60 Rockwell C hardness. It can be used for slotting, side milling and contouring applications. The ball nose produces a smooth milling finish. The high-speed steel construction provides wear resistance and allows milling speeds up to 100 SFM.
Catalog No. 50-275-3349