Learn More
Grainger Ball End Mill Single End 4.00mm Carbide
Supplier:  Grainger 14K756
Encompass_Preferred
The Ball End Mill from Ball is a high-speed steel end mill with a ball nose end. It has a 1/2" shank diameter, a 2" overall length, and a 1/2" cutting diameter. The end mill has a TiN coating for increased hardness and lubricity. It is designed for general purpose milling in materials like aluminum, brass, bronze, and copper. The end mill provides accurate results and smooth finishes at high RPMs. With its durable construction, it is well-suited for high production milling applications.
Catalog No. 50-275-2728