Learn More
Grainger Ball End Mill Single End 4.00mm Carbide
Supplier:  Grainger 14K385
Encompass_Preferred
The Micro 100 Ball End Mill is a 2 flute carbide end mill with a ball end. It has a 1/4" shank diameter and is available in multiple diameters from 1/8" to 1/2". The ball end allows milling of 3D contoured surfaces. Carbide provides hardness, wear resistance, and heat resistance. It can machine steels, stainless steels, cast iron, non-ferrous metals, and non-metallic materials. This versatile end mill is well-suited for complex 3D contouring and finishing in mold, die, and prototype work.
Catalog No. 50-274-9923