Learn More
Grainger PARKER Ball Valve Full
Supplier:  Grainger 5UMU9
Encompass_Preferred
The Parker 5UMU9 is a 1/4 inch full port brass ball valve with a 316 stainless steel ball and stem. It has a pressure rating of 6000 psi and temperature rating of -65 to 450°F. The valve has female NPT threaded ends and comes with a vinyl grip handle for manual operation. This valve can be used for a variety of general purpose fluid and gas applications. The full port design allows for minimal pressure drop.
Catalog No. NC2388634