Learn More
Medchemexpress LLC Balofloxacin | 127294-70-6 | MFCD00864925 | 98.3% | 10 G
SDP
Supplier:  Medchemexpress LLC HYB015910G
Encompass_Preferred
Balofloxacin (Q-35) is an orally active fluoroquinolone antibiotic known for its broad-spectrum antibacterial activity. It is effective against gram-negative, gram-positive, and anaerobic bacteria, making it suitable for research applications related to respiratory, intestinal, and urinary tract infections.
  • Orally active fluoroquinolone antibiotic.
  • Exhibits broad-spectrum antibacterial activity.
  • Effective against gram-negative, gram-positive, and anaerobic bacteria.
  • Used in research for respiratory, intestinal, and urinary tract infections.
  • Decreases growth of *S. aureus* adhered to human corneal epithelial cells.
  • Inhibits *S. aureus* with a MIC of 0.125 mg/mL.
  • Does not induce toxicity on human corneal epithelial cells at low concentrations.
  • Reduces viable *M. pneumonia* counts in hamster lungs.
  • Eye drops are effective in *S. aureus* bacterial keratitis.
Catalog No. 50-004-06239