Learn More
Medchemexpress LLC Balovaptan | 1228088-30-9 | 99.9% | 409.91 | 1 ML

Supplier: Medchemexpress LLC HY10902410MM/1ML
Balovaptan (RG7314) is an orally available, selective brain-penetrant vasopressin 1a (hV1a) receptor antagonist, with Kᵢs of 1 and 39 nM for human (hV1a) and mouse (mV1a) receptors. It is used for the research of autism and shows >30000-fold selectivity for hV1a over hV2 receptors, and 9891-fold selectivity over hOTR (human oxytocin receptor).
- Orally available
- Selective brain-penetrant vasopressin 1a (hV1a) receptor antagonist
- Used for the research of autism
- Appearance: Solid
- Color: White to off-white
- Shipping at room temperature in continental US; may vary elsewhere
- Storage as powder at -20°C for 3 years, 4°C for 2 years
- Storage in solvent at -80°C for 2 years, -20°C for 1 year
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.