Learn More
Medchemexpress LLC Balovaptan | 1228088-30-9 | 99.9% | 409.91 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY109024100MG
Encompass_Preferred
Balovaptan (RG7314) is an orally available, selective brain-penetrant vasopressin 1a (hV1a) receptor antagonist. It is characterized by its high selectivity for hV1a receptors and is commonly used in research related to autism.
  • Orally available
  • Selective brain-penetrant vasopressin 1a (hV1a) receptor antagonist
  • High selectivity for hV1a receptors over hV2 and hOTR
  • Utilized in autism research
Catalog No. 50-003-45926