Learn More
Medchemexpress LLC Balovaptan | 1228088-30-9 | 99.9% | 409.91 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY10902450MG
Encompass_Preferred
Balovaptan is an orally available, selective brain-penetrant vasopressin 1a (hV1a) receptor antagonist. It is utilized in autism research and exhibits high selectivity for its target receptors.
  • Orally available
  • Selective brain-penetrant vasopressin 1a (hV1a) receptor antagonist
  • Ki values of 1 nM for human (hV1a) and 39 nM for mouse (mV1a) receptors
  • Greater than 30000-fold selectivity for hV1a over hV2 receptors
  • 9891-fold selectivity over human oxytocin receptor (hOTR)
Catalog No. 50-003-44520