Learn More
Medchemexpress LLC BAM7 | 331244-89-4 | C21H19N5O2S | 50 MG
SDP
Supplier:  Medchemexpress LLC HY1534150MG
Encompass_Preferred
BAM7 is a direct and selective activator of proapoptotic BAX with an IC50 of 3.3 μM.
  • Selective for the BH3-binding site on BAX
  • Activates BAX and BAX-dependent cell death
  • Stable under recommended storage conditions
  • For research use only
Catalog No. 50-003-72649