Learn More
Medchemexpress LLC BAMB-4 (ITPKA-IN-C14) | 891025-25-5 | 99.99% | 252.27 | 1ML
SDP
Supplier:  Medchemexpress LLC HY1669410MM/1ML
Encompass_Preferred
BAMB-4 (ITPKA-IN-C14) is a specific and membrane-permeable ITPKA inhibitor. It exhibits high stability and membrane permeability. It inhibits the inositol-1,4,5-trisphosphate (InsP3) kinase activity of inositol-1,4,5-trisphosphate-3-kinase A (ITPKA) with an IC50 value of 20 μM. BAMB-4 can be used for research related to the metastasis of lung cancer. It is for research use only and not sold to patients.
  • Specific and membrane-permeable ITPKA inhibitor.
  • High stability and membrane permeability.
  • Inhibits inositol-1,4,5-trisphosphate (InsP3) kinase activity of ITPKA with an IC50 of 20 μM.
  • IC50 for ITPKA is 37 μM.
  • High cellular uptake.
  • Acts as a mixed-type inhibitor with respect to ATP and InsP3, affecting binding of both substrates.
  • Used for research of metastasis of lung cancer.
Catalog No. 50-003-94205