Learn More
Medchemexpress LLC Bambermycin | 11015-37-5 | 99.9% | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY126387
Encompass_Preferred
Moenomycin complex is a class of antibacterial agents and PBP inhibitors, with Kd values ranging from 94 nM to 1690 nM against various bacterial PBPs. It inhibits bacterial growth by blocking the transglycosylase activity of class A penicillin-binding proteins (PBPs) and is used in the research of bacterial infections.
  • Class of antibacterial agents and PBP inhibitors.
  • Kd values ranging from 94 nM to 1690 nM against various bacterial PBPs.
  • Inhibits bacterial growth by blocking the transglycosylase activity of class A penicillin-binding proteins (PBPs).
  • Used in the research of bacterial infections.
  • Binds to full-length class A PBPs from 16 bacterial species.
Catalog No. 50-004-40066