Learn More
DYNAMISM INC Bambu Lab PETG HF Black - 1kg
SDP
Supplier:  DYNAMISM INC 33102
Encompass
Key Features:Smooth, soft textureAMS compatibleIncludes reusable spool Please Note: Requires drying to achieve optimal print quality.
Catalog No. NC3595023