Learn More
Cole-Parmer Pomona Electronics 22495 2 ft (0.6 M) 50 Ohm Cable 0.195" (5mm) O.D. RG58C/U
Supplier:  Cole-Parmer 2000745
Encompass
This BNC cable with molded strain relief is a 50 RG58C/U cable and ideal for most test and measurement applications. It is available for sale solely in the US and Canada.
Catalog No. 50-993-195