Learn More
Med Vet International Miltex Vantage Universal Bandage And Utility Scissors, 7.5in., Yellow Handle
Supplier:  Med Vet International MILV951030
Encompass_Preferred
The Miltex Vantage V95-1030 universal bandage and utility scissors are 7.5" in length and have yellow handles. They are designed to cut bandage and gauze, cast material, metal sheets, and tough clothing material. Autoclavable to 143C or 290F for easy sterilization.
Catalog No. 50-283-2036