Learn More
Sigma Aldrich Fine Chemicals Biosciences Nα-Benzoyl-DL-arginine 4-nitroanilide hydrochloride, 911-77-3, MFCD00012846, 1G
Supplier:  Sigma Aldrich Fine Chemicals Biosciences B48751G
Encompass
Molecular Formula: C19H22N6O4 · HCl, Molecular Weight: 434.88, Assay: ≥98%
Catalog No. NC0145386