Learn More
Medchemexpress LLC Bapta-am | 126150-97-8 | MFCD00036696 | ≥90.0% | 764.68 g·mol⁻¹ | C34H40N2O18 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY100545100MG
Encompass_Preferred
BAPTA-AM is a membrane-permeable calcium chelator commonly used to buffer intracellular Ca2+ levels in cell-based assays and electrophysiology studies. Supplied as a powder, it is typically dissolved in DMSO to prepare concentrated stock solutions for cellular loading.
Catalog No. 50-002-11930