Learn More
Apexbio Technology LLC BAPTA-AM, 50mg. Cas: 126150-97-8 MFCD: MFCD00036696. calcium chelator, selective and membrane permeable
SDP
Supplier:  Apexbio Technology LLC B475850
Encompass_Preferred
BAPTA-AM is a selective calcium chelator [1]. Ca2+ is one of the most ubiquitous and versatile intracellular signaling molecules that control numerous cellular processes such as neurotransmitter release, contraction of all muscle cell types and fertilization [4]. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5360