Learn More
Abcam BAPTA, e x tracellular calcium chelator, 250MG
Supplier:  Abcam AB144924250MG
Encompass_Preferred
MW 476.4 Da. Selective, membrane impermeable extracellular calcium chelator. Binds Ca²+ ions in a 2:1 ratio. Shows 105-fold greater affinity for Ca²+ than Mg²+. BAPTA-AM (ab120503) and BAPTA sodium salt (ab120449) analog.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5460