Learn More
Medchemexpress LLC Bapta tetrapotassium | 73630-08-7 | MFCD00036255 | 99.5% | 628.79 | C22H20K4N2O10 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY100168B100MG
Encompass_Preferred
BAPTA tetrapotassium is a selective intracellular calcium chelator used to buffer Ca2+ concentrations and study calcium-dependent signaling pathways. The tetrapotassium salt is water-soluble and commonly used in cell biology, neuroscience, and biochemical assays; it can also inhibit phospholipase C independently of chelation.
  • Selective chelator for calcium ions.
  • High selectivity against magnesium ions.
  • Water-soluble tetrapotassium salt suitable for intracellular buffering.
  • Useful in cell biology, neuroscience, and biochemical assays.
  • Available in small laboratory quantities for experimental use.
Catalog No. 50-002-15740