Learn More
Selleck Chemical LLC BAPTA tetrapotassium salt E2691-25MG
SDP
Supplier:  Selleck Chemical LLC E269125MG
Encompass_Preferred
BAPTA tetrapotassium salt (BAPTA tetrapotassium) is a non-permeable selective extracellular calcium chelator with 105-fold greater affinity for Ca2 than Mg2 which is a valuable tool to study the role of calcium in cell signaling
Catalog No. 50-000-39895