Learn More
TARGETMOL CHEMICALS INC BAR501 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T40835MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. BAR501 is an effective and specific GPBAR1 agonist (EC50 1 uM). purity: 99%
Catalog No. 50-257-6813