Learn More
Medchemexpress LLC HY-101274 5mg Medchemexpress, BAR501 CAS:1632118-69-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1012745MG
Encompass_Preferred
Medchemexpress, HY-101274 5mg BAR501 CAS:1632118-69-4 BAR501 is a potent and selective agonist of GPBAR1 with an EC 50 of 1 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2084