Learn More
TARGETMOL CHEMICALS INC Barasertib 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T1437110MG
Encompass_Preferred
Also available in 1 mL, 2 mg, 5 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. AZD1152 is a pro-drug of Barasertib (AZD1152)-hQPA. Which is a highly selective Aurora B inhibitor with IC50 of 0.37 nM in a cell-free assay. Purity 99.63%
Catalog No. 50-287-5873