Learn More
Cole-Parmer Hose Barb Fitting, Straight Reducer, Medical Nylon, Cleanroom Packed, 1/4" ID x 5/32" ID; 10/Pk
Supplier:  Cole-Parmer 5010893
Encompass_Preferred
Use these high-quality PVC-free fittings in biomedical, pharmaceutical, electronic, and other critical-use applications. Fittings are manufactured, double bagged, and sealed in an ISO Class 7 cleanroom to ensure that the integrity is not compromised. An ISO Class 7 cleanroom utilizes HEPA filtration systems to maintain air cleanliness levels of less than 10,000 particles of 0.5 µm or larger per cubic foot.
Easily connect two different size tubing with straight reducers. A single barb on each end of the fitting creates a tight seal to keep the tubing in place. These medical nylon fittings are made from the same resin as natural nylon but with FDA-compliant white coloring. They offer good tensile strength, stiffness, and impact resistance and are resistant to fuels and lubricants. Not recommended for use with phenols, strong acids, and oxidizing agents.
Catalog No. 50-255-8334