Learn More
Medchemexpress LLC HY-119706 1mg Medchemexpress, Barbadin CAS:356568-70-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1197061MG
Encompass_Preferred
Medchemexpress, HY-119706 1mg Barbadin CAS:356568-70-2 Barbadin is a novel and selective β-arrestin/β2-adaptin interaction inhibitor, has IC50 values of 19.1 μM for β-arrestin1 and 15.6 μM for β-arrestin2. Barbadin blocks agonist-promoted endocytosis of the prototypical β2-adrenergic, V2-vasopressin and angiotensin-II type-1 receptors. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9544