Learn More
Accuflow Systems BarbLock® Ultra-Secure Retainer
Supplier: Accuflow Systems BL135375
BarbLock® Ultra-Secure Retainers are designed for demanding connective applications, offering a full 360° radial crimp behind the barb and 360° of compression directly over the barb's ridge for retaining flexible tubing and hose onto barbed fittings. They do not use adhesives or solvents, making them ideal for systems transferring delicate liquids or gases, and are quick and easy to assemble.
- Replaces inconsistent and leak-prone cable ties and traditional clamps.
- Compatible with all flexible tubing materials.
- Provides 360° radial crimp behind the barb.
- Supplies 360° compression over the barb and full dynamic retention.
- Eliminates solvent bonding, ultraviolet bonding and toxic adhesives.
- Constructed with USP Class VI materials.
- Quick and easy to assemble.