Learn More
Cayman Chemical BARD1 BRCT domaInshumnre 100ug
SDP
Supplier:  Cayman Chemical 14166100 UG
Encompass_Preferred
Source: Recombinant N-terminal GST-tagged protein expressed in E. coli • Mr: 52.4 kDa
Catalog No. 50-295-2925