Learn More
Medchemexpress LLC Baricitinib | 1187594-09-7 | C16H17N7O2S | 500 MG

Supplier: Medchemexpress LLC HY15315500MG
Baricitinib is a selective and orally bioavailable inhibitor of JAK1 and JAK2. This compound is intended for research applications, facilitating studies in inflammation and immunology.
- Selective and orally bioavailable JAK1 and JAK2 inhibitor
- IC50s of 5.9 nM for JAK1 and 5.7 nM for JAK2
- Molecular weight: 371.42
- Appearance: Solid
- Color: White to off-white
- Melting/freezing point: 212-215°C
- Relative density: 1.56 g/cm³
- Inhibits IL-6 induced MCP-1 production in peripheral blood mononuclear cells (IC50 40 nM)
- Inhibits pSTAT3 stimulated by IL-23 in isolated naive T-cells (IC50 20 nM)
- Prevents production of IL-17 and IL-22 by Th17 cells (IC50 50 nM)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.