Learn More
Medchemexpress LLC Alpha-naphtholphthalein | 596-01-0 | 99.7% | 418.44 | C28H18O4 | 1 G

Supplier: Medchemexpress LLC HYW0879131G
Alpha-naphtholphthalein is a pH indicator dye used in acid-base titrations and pH sensing. It is an off-white to light yellow solid with molecular formula C28H18O4 and molecular weight 418.44 g/mol. Typical applications include analytical chemistry and laboratory pH measurements.
- pH indicator that changes from light yellow at pH 6 to green/blue at higher pH.
- High purity (99.71%).
- Solid, off-white to light yellow appearance.
- Available as 1 g laboratory reagent.
- Storage: keep sealed at 4°C; in solvent store at -80°C for up to 6 months or -20°C for up to 1 month.
- Suitable for titration and pH sensing applications.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.