Learn More
Strem, An Ascensus Company CAS# 6865-35-6. 250g. Barium stearate, tech. gr.. MFCD00050711
SDP
Supplier:  Strem, An Ascensus Company 935644250G
Encompass
CAS# 6865-35-6. 250g. Barium stearate, tech. gr.. MFCD00050711. Molecular Weight: 704.30. Molecular Formula: Ba(O2C18H35)2. Color/form: white pwdr. Strem# 93-5644. http://www.strem.com/catalog/v/93-5644/
Catalog No. 50-901-15812