Learn More
BURKLE INC Solvent pump foot operated with discharge tube, 37.40 in (95 cm)
Supplier:  BURKLE INC 56031100
Encompass_Preferred
Designed for pouring flammable liquids, this solvent pump features all medium-contact parts made from durable stainless steel AISI 304 (1.4301) and PTFE. For added safety, the Bürkle anti-static set can ground the pump, preventing electrostatic charges during dispensing.
The pump and container can be hermetically sealed thanks to the barrel screw joint and stopcock, effectively preventing hazardous vapors from escaping after use—protecting both health and the environment.
Built for durability and practicality, this industrial-grade solvent pump operates on the overpressure principle. By using the foot actuator, excess pressure builds inside the container, pushing the liquid out smoothly. The hands-free foot operation makes dispensing easier and safer. Additionally, the air hose connects and disconnects quickly and securely via a convenient clip connection.
Catalog No. 50-000-25715