Learn More
Cole-Parmer High Sensitivity Noncontact Probe
Supplier:  Cole-Parmer 2002096
Encompass_Preferred
Mag-Probe is a magnetic sensing instrument used to trouble-shoot AC and DC powered solenoids, solenoid valves, relays, mercury wetted relays, heat controllers and test coils used in robotics even automotive alternators, voltage regulators and electronic ignitions systems. Unit is ideal for testing the largest to sub-miniature solenoids, relays and most reed relays and all automotive applications.
Catalog No. NC0454755